Invitrogen
Manufacturer Code:PA561593
Catalog # PIPA561593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CABYR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2); Calcium-binding protein 86; Calcium-binding tyrosine phosphorylation-regulated protein; Calcium-binding tyrosine phosphorylation-regulated protein Calcium-binding protein 86 Cancer/testis antigen 88 CT88 Fibrousheathin II Fibrousheathin-2 FSP-2 Testis-specific calcium-binding protein CBP86 CBP86 FSP2; calcium-binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2); Cancer/testis antigen 88; CT88; fibrousheathin 2; Fibrousheathin II; Fibrousheathin-2; FSP-2; testis tissue sperm-binding protein Li 84P; Testis-specific calcium-binding protein CBP86
Gene Aliases: CABYR; CABYRa; CABYRc; CABYRc/d; CABYRe; CBP86; CT88; FSP-2; FSP2
UniProt ID: (Human) O75952
Entrez Gene ID: (Human) 26256
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.