Novus Biologicals
Manufacturer Code:NBP272071UV
Catalog # NBP272071UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9A7 |
Immunogen | Full length recombinant protein of human CABP4 (NP_660201) produced in E.coli. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CaBP4; CaBP4 calcium binding protein 4 CSNB2Bcalcium-binding protein 4; Calcium-binding protein 4
Gene Aliases: CABP4; CRSD; CSNB2B
UniProt ID: (Human) P57796
Entrez Gene ID: (Human) 57010
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.