Invitrogen
Manufacturer Code:PA577382
Catalog # PIPA577382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RTYGPLLNRMFGKDR corresponding to amino acid residues 105-120 of mouse Calcium-Binding Protein 4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CaBP4; Calcium-Binding Protein 4
Gene Aliases: 2410038D05Rik; Cabp4
UniProt ID: (Mouse) Q8VHC5
Entrez Gene ID: (Mouse) 73660, (Rat) 365394
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.