Invitrogen
Manufacturer Code:PA570636
Catalog # PIPA570636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of rat CABP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CaBP4; cabp4a; Calcium-binding protein 4; Calcium-binding protein 4 CaBP4
Gene Aliases: 2410038D05Rik; CABP4; CRSD; CSNB2B; si:ch73-56f16.1
UniProt ID: (Bovine) Q8HZJ4, (Human) P57796, (Mouse) Q8VHC5
Entrez Gene ID: (Bovine) 281656, (Dog) 483694, (Human) 57010, (Horse) 100059382, (Mouse) 73660, (Guinea pig) 100722978, (Rat) 365394, (Zebrafish) 797905
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.