Invitrogen
Manufacturer Code:PA5103682
Catalog # PIPA5103682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CABP4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410038D05Rik CABP4 calcium binding protein 4 Calcium-binding protein 4 CRSD CSNB2B; CaBP4; Calcium-binding protein 4
Gene Aliases: 2410038D05Rik; CABP4; CRSD; CSNB2B
UniProt ID: (Human) P57796, (Mouse) Q8VHC5
Entrez Gene ID: (Human) 57010, (Mouse) 73660
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.