Invitrogen
Manufacturer Code:PA529172
Catalog # PIPA529172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 341 of Human CAB39 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium binding protein, 39 kDa; Calcium-binding protein 39; MO25 MO25alpha calcium-binding protein 39; MO25alpha; Protein Mo25
Gene Aliases: AA408805; AA960512; C78372; CAB39; CGI-66; MO25; MO25alpha
UniProt ID: (Human) Q9Y376, (Mouse) Q06138
Entrez Gene ID: (Human) 51719, (Mouse) 12283
Molecular Function:
annexin
calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.