Proteintech
Manufacturer Code:662431IG150UL
Catalog # 50-173-6676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G4G5 |
Immunogen | CA9 Fusion Protein Ag1540 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CA IX CA9 CAIX Carbonate dehydratase IX Carbonic anhydrase 9 carbonic anhydrase IX G250 Membrane antigen MN MN P54/58N pMW1 RCC associated antigen G250; CA-IX; Carbonate dehydratase IX; Carbonic anhydrase 9; Carbonic anhydrase IX; carbonic dehydratase; Membrane antigen MN; Membrane antigen MN homolog; P54/58N; pMW1; RCC-associated antigen G250; RCC-associated protein G250; Renal cell carcinoma-associated antigen G250
Gene Aliases: CA9; CAIX; Car9; G250; MN; MN/CA9
UniProt ID: (Human) Q16790, (Mouse) Q8VHB5
Entrez Gene ID: (Pig) 100152792, (Human) 768, (Mouse) 230099
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.