Invitrogen
Manufacturer Code:PA570469
Catalog # PIPA570469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human CA7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CA-VII; Carbonate dehydratase VII; Carbonic anhydrase 7; Carbonic anhydrase VII; carbonic dehydratase VII; CAVII CA-VII carbonate dehydratase VII carbonic anhydrase 7 carbonic dehydratase VII
Gene Aliases: AV343731; CA7; Car7; CAVII
UniProt ID: (Human) P43166, (Mouse) Q9ERQ8
Entrez Gene ID: (Dog) 489772, (Rabbit) 100350484, (Bovine) 520403, (Human) 766, (Horse) 100065583, (Guinea pig) 100715522, (Mouse) 12354
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.