Invitrogen
Manufacturer Code:PA5103703
Catalog # PIPA5103703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CA7 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AV343731 CA7 Car7 carbonate dehydratase VII Carbonic anhydrase 7 carbonic anhydrase VII carbonic dehydratase VII CAVII CA-VII; CA-VII; Carbonate dehydratase VII; Carbonic anhydrase 7; Carbonic anhydrase VII; carbonic dehydratase VII
Gene Aliases: AV343731; CA7; Car7; CAVII
UniProt ID: (Human) P43166, (Mouse) Q9ERQ8
Entrez Gene ID: (Human) 766, (Mouse) 12354, (Rat) 291819
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.