Proteintech
Manufacturer Code:154351AP150UL
Catalog # 50-173-0225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CA11 Fusion Protein Ag7712 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CA RP II CA RP XI CA XI CA11 carbonic anhydrase XI CARP 2 CARP XI CARP2 CARPX1; CA-RP II; CA-RP XI; CA-XI; carbonic anhydrase XI; Carbonic anhydrase-related protein 11; Carbonic anhydrase-related protein 2; carbonic anhydrase-related protein XI; carbonic anhydrase-related XI protein; CARP XI; CARP-2
Gene Aliases: CA11; CAR-XI; Car11; CARP2; CARPX1; UNQ211/PRO237
UniProt ID: (Human) O75493, (Mouse) O70354
Entrez Gene ID: (Human) 770, (Rat) 308588, (Mouse) 12348
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.