OriGene
Manufacturer Code:TA811304
Catalog # 50-168-3969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B12 |
Immunogen | Full length human Recombit protein of human C9ORF89 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bcl10-interacting CARD protein; Bcl10-interacting protein with CARD; BinCARD; BinCARD C9orf89; caspase recruitment domain family, member 19; Caspase recruitment domain-containing protein 19
Gene Aliases: BinCARD; C9orf89; CARD19
UniProt ID: (Human) Q96LW7
Entrez Gene ID: (Human) 84270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.