Proteintech
Manufacturer Code:226371AP150UL
Catalog # 50-554-580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C9orf72 Fusion Protein Ag18326 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C9orf72 C9RANT RP11 27J8.2; Guanine nucleotide exchange C9orf72; Guanine nucleotide exchange C9orf72 homolog; protein C9orf72 homolog
Gene Aliases: AI840585; ALSFTD; C9orf72; FTDALS; FTDALS1
UniProt ID: (Mouse) Q6DFW0, (Human) Q96LT7, (Rat) Q66HC3
Entrez Gene ID: (Mouse) 73205, (Human) 203228, (Rat) 313155
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.