Novus Biologicals
Manufacturer Code:NBP15946220UL
Catalog # NBP15946220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to C9ORF46 The peptide sequence was selected from the middle region of C9ORF46. Peptide sequence AIKKKKPAFLVPIVPLSFILTYQYDLGYGTLLERMKGEAEDILETEKSKL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5033414D02Rik; AD025 C9orf46 chromosome 9 open reading frame 465033414D02Rik FLJ14688 FLJ39176 MDS030 plasminogen receptor C-terminal lysine transmembrane protein Plg-R(KT) PLG-RKT transmembrane protein C9orf46; Plasminogen receptor (KT); plasminogen receptor, C-terminal lysine transmembrane protein; Plg-R(KT); transmembrane protein C9orf46
Gene Aliases: AD025; C9orf46; MDS030; Plg-R(KT); PLG-RKT; PLGRKT
UniProt ID: (Human) Q9HBL7
Entrez Gene ID: (Human) 55848
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.