Abnova
Manufacturer Code:H00414328A01
Catalog # 89-107-803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 41 a.a. to 141 a.a) partial recombinant protein with GST tag.,C9orf103 (NP_001001551 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant C9orf103.
For Research Use Only
Protein Aliases: bA522I20.2; glucokinase-like protein; Gluconate kinase; gluconokinase-like protein; idnK, gluconokinase homolog; Probable gluconokinase
Gene Aliases: bA522I20.2; C9orf103; hGntK; IDNK
UniProt ID: (Human) Q5T6J7
Entrez Gene ID: (Human) 414328
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.