Invitrogen
Manufacturer Code:PA571292
Catalog # PIPA571292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human C7orf72 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 9 C7orf72 homolog; Post-meiotic spermatogenesis protein 1; Spermatogenesis-associated protein 48; uncharacterized protein C7orf72 homolog
Gene Aliases: C18H7orf72; C4H7orf72; C9H7orf72; Pms1; Spata48
UniProt ID: (Mouse) Q5NC83
Entrez Gene ID: (Mouse) 73862, (Human) 100130988, (Rat) 289778, (Horse) 100630285, (Pig) 100625163, (Dog) 475861, (Guinea pig) 100734317
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.