Proteintech
Manufacturer Code:249571AP150UL
Catalog # 50-173-4643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C6orf182 Fusion Protein Ag21054 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf182 Centrosomal protein 57kDa-like protein 1 Cep57 related protein CEP57L1 Cep57R; centrosomal protein 57kDa-like 1; Centrosomal protein 57kDa-like protein 1; Centrosomal protein CEP57L1; Centrosomal protein of 57 kDa-related protein; Cep57-related protein; Cep57R
Gene Aliases: 2410017P07Rik; 4930484D11Rik; AI607327; bA487F23.2; C6orf182; CEP57L1; CEP57R
UniProt ID: (Human) Q8IYX8, (Mouse) Q8VDS7
Entrez Gene ID: (Human) 285753, (Mouse) 103268
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.