Invitrogen
Manufacturer Code:PA553057
Catalog # PIPA553057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human C6orf170 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA301B7.2 (novel protein); broad-minded homolog; BROMI C6orf171 bA301B7.2 bA57L9.1 dJ310J6.1 bA301B7.2 (novel protein) dJ310J6.1 (novel protein) protein broad-minded; dJ310J6.1 (novel protein); Protein broad-minded; TBC1 domain family member 32; TBC1 domain family, member 32
Gene Aliases: bA301B7.2; bA57L9.1; BROMI; C6orf170; C6orf171; dJ310J6.1; TBC1D32
UniProt ID: (Human) Q5SZD6
Entrez Gene ID: (Human) 221322
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.