Invitrogen
Manufacturer Code:PA526035
Catalog # PIPA526035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 112-139 amino acids from the N-terminal region of human C6orf170 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA301B7.2 (novel protein); broad-minded homolog; BROMI C6orf171 bA301B7.2 bA57L9.1 dJ310J6.1 bA301B7.2 (novel protein) dJ310J6.1 (novel protein) protein broad-minded; dJ310J6.1 (novel protein); Protein broad-minded; TBC1 domain family member 32; TBC1 domain family, member 32
Gene Aliases: bA301B7.2; bA57L9.1; BROMI; C6orf170; C6orf171; dJ310J6.1; TBC1D32
UniProt ID: (Human) Q5SZD6
Entrez Gene ID: (Human) 221322
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.