Abnova
Manufacturer Code:PAB28228
Catalog # 89-129-733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of recombinant C6orf170. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant C6orf170.
For Research Use Only
Protein Aliases: bA301B7.2; bA301B7.2 (novel protein); bA57L9.1; broad-minded homolog; C6orf171; dJ310J6.1; dJ310J6.1 (novel protein); FLJ30899; FLJ34235; Protein broad-minded; TBC1 domain family member 32; TBC1 domain family, member 32
Gene Aliases: bA301B7.2; bA57L9.1; BROMI; C6orf170; C6orf171; dJ310J6.1; TBC1D32
UniProt ID: (Human) Q5SZD6
Entrez Gene ID: (Human) 221322
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.