Invitrogen
Manufacturer Code:PA5113022
Catalog # PIPA5113022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human MRN complex-interacting protein (1-200AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C5orf45 chromosome 5 open reading frame 45 MRN complex-interacting protein MRN-interacting protein MRNIP UPF0544 protein C5orf45; MRN complex-interacting protein; MRN-interacting protein; UPF0544 protein C5orf45 homolog
Gene Aliases: 2900016G09Rik; AI851514; AW475975; C5orf45; MRNIP
UniProt ID: (Mouse) Q9D1F5, (Human) E9PAK6
Entrez Gene ID: (Mouse) 68067, (Human) 51149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.