Proteintech
Manufacturer Code:213161AP150UL
Catalog # 50-173-3365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C5aR Fusion Protein Ag15968 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5A C5a R C5aR C5AR1 C5R1 CD88; C5a ligand; C5a-R; C5aR; CD88; complement C5a receptor; complement component 5 receptor 1; complement component 5, receptor 1
Gene Aliases: C5A; C5AR; C5AR1; C5R1; CD88; D7Msu1
UniProt ID: (Human) P21730, (Mouse) P30993, (Rat) P97520
Entrez Gene ID: (Human) 728, (Mouse) 12273, (Rat) 113959
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.