Abnova
Manufacturer Code:H00153222K
Catalog # 89-209-432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human C5ORF41 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human C5orf41 peptide using ARM Technology.
For Research Use Only
Protein Aliases: adult retina protein; CREB3 regulatory factor; DKFZp313F2319 DKFZp686G2059 LRF; LRF; Luman recruitment factor; luman-recruiting factor; luman/CREB3 recruitment factor; UPF0474 protein C5orf41
Gene Aliases: C5orf41; CREBRF; LRF
UniProt ID: (Human) Q8IUR6
Entrez Gene ID: (Human) 153222
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.