R&D Systems
Manufacturer Code:MAB4729
Catalog # MAB4729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 468705 |
Immunogen | Chinese hamster ovary cell line transfected with mouse C5L2 Met1-Val344 Accession # Q8BW93 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C5a anaphylatoxin chemotactic receptor 2; C5a anaphylatoxin chemotactic receptor C5L2; C5a anaphylatoxin chemotactic receptor C5L2 C5L2G-protein coupled receptor 77 G protein-coupled receptor 77 G protein-coupled receptor C5L2 GPF77 GPR77; Complement component 5a receptor 2; G protein-coupled receptor 77; G protein-coupled receptor C5L2; G-protein coupled receptor 77
Gene Aliases: C5AR2; C5L2; GPF77; GPR77
UniProt ID: (Human) Q9P296
Entrez Gene ID: (Human) 27202
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.