Proteintech
Manufacturer Code:250361AP150UL
Catalog # 50-173-4685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C4orf29 Fusion Protein Ag19120 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abhydrolase domain-containing protein 18; Alpha/beta hydrolase domain-containing protein 18; C4orf29; Protein ABHD18; uncharacterized protein C4orf29; uncharacterized protein C4orf29 homolog
Gene Aliases: 2310068E01Rik; 4732404H23; Abhd18; AI645591; C4orf29
UniProt ID: (Mouse) Q8C1A9
Entrez Gene ID: (Mouse) 269423, (Human) 80167
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.