Proteintech
Manufacturer Code:118191AP150UL
Catalog # 50-172-7480
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C4BPA Fusion Protein Ag2387 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C4b-binding protein alpha chain; C4bp; C4BP C4BPA Proline rich protein PRP; Proline-rich protein; PRP
Gene Aliases: C4BP; C4BPA; PRP
UniProt ID: (Human) P04003
Entrez Gene ID: (Human) 722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.