Novus Biologicals
Manufacturer Code:NBP158983
Catalog # NBP158983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to C4BPB (complement component 4 binding protein beta) The peptide sequence was selected from the N terminal of C4BPB. Peptide sequence CCLMVAWRVSASDAEHCPELPPVDNSIFVAKEVEGQILGTYVCIKGYHLV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C4b binding protein beta chain C4b-binding protein beta chain C4BP complement component 4 binding protein beta complement component 4 binding protein beta chain complement component 4-binding protein beta; C4b-binding protein beta chain
Gene Aliases: C4BP; C4BPB
UniProt ID: (Human) P20851
Entrez Gene ID: (Human) 725
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.