Abnova
Manufacturer Code:PAB31570
Catalog # 89-998-511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human C3orf39. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGO61; Extracellular O-linked N-acetylglucosamine transferase-like; FLJ14566; glycosyltransferase-like domain containing 2; Glycosyltransferase-like domain-containing protein 2; POMGnT2; Protein O-linked-mannose beta-1,4-N-acetylglucosaminyltransferase 2
Gene Aliases: AGO61; C3orf39; EOGTL; GTDC2; MDDGA8; POMGNT2
UniProt ID: (Human) Q8NAT1
Entrez Gene ID: (Human) 84892
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.