Invitrogen
Manufacturer Code:PA543095
Catalog # PIPA543095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human C3orf31 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDP-DAG synthase; CDP-diacylglycerol synthase; Mitochondrial translocator assembly and maintenance protein 41 homolog; MMP37-like protein, mitochondrial; Phosphatidate cytidylyltransferase, mitochondrial; TAM41; TAM41, mitochondrial translocator assembly and maintenance protein, homolog
Gene Aliases: 1500001M20Rik; C20H3orf31; C22H3orf31; C3orf31; RAM41; TAM41; TAMM41
UniProt ID: (Bovine) Q32L81, (Human) Q96BW9, (Rat) D3ZKT0
Entrez Gene ID: (Pig) 100524902, (Rabbit) 100348148, (Bovine) 514330, (Horse) 100051402, (Human) 132001, (Dog) 476532, (Guinea pig) 100725702, (Mouse) 68971, (Rat) 362419
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.