Invitrogen
Manufacturer Code:PA5113300
Catalog # PIPA5113300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Phosphatidate cytidylyltransferase mitochondrial protein (161-452AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500001M20Rik BOS_21154 C20H3orf31 C22H3orf31 C3orf31 CDP-DAG synthase CDP-diacylglycerol synthase LOC100348148 Mitochondrial translocator assembly and maintenance protein 41 homolog MMP37-like protein mitochondrial MMP37-like protein mitochondrial-like Phosphatidate cytidylyltransferase mitochondrial RAM41 TAM41 TAM41 mitochondrial translocator assembly and maintenance homolog TAM41 mitochondrial translocator assembly and maintenance protein homolog TAM41 mitochondrial translocator assembly and maintenance protein homolog (S. cerevisiae) TAMM41; CDP-DAG synthase; CDP-diacylglycerol synthase; Mitochondrial translocator assembly and maintenance protein 41 homolog; MMP37-like protein, mitochondrial; Phosphatidate cytidylyltransferase, mitochondrial; TAM41; TAM41, mitochondrial translocator assembly and maintenance protein, homolog
Gene Aliases: 1500001M20Rik; C3orf31; RAM41; TAM41; TAMM41
UniProt ID: (Human) Q96BW9
Entrez Gene ID: (Human) 132001, (Mouse) 68971
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.