Invitrogen
Manufacturer Code:MA517473
Catalog # PIMA517473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 74 |
Immunogen | RBL-2H3 transfectants expressing rat C3aR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C3a anaphylatoxin chemotactic receptor; C3a-R; C3AR; C3AR1 AZ3B C3AR C3R1 C3a receptor C3a-R HNFAG09 Anaphylatoxin c3a receptor C3a anaphylatoxin receptor
Gene Aliases: C3ar1
UniProt ID: (Rat) O55197
Entrez Gene ID: (Rat) 84007
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.