Invitrogen
Manufacturer Code:PA5109467
Catalog # PIPA5109467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 161-337 of human C3AR1 (NP_004045.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anaphylatoxin C3a receptor; AZ3B C3a anaphylatoxin chemotactic receptor C3a receptor C3AR C3ar1 C3R1 Complement component 3a receptor 1 HNFAG09; C3a anaphylatoxin chemotactic receptor; C3a-R; C3AR; complement component 3 receptor 1; Complement component 3a receptor 1
Gene Aliases: AZ3B; C3AR; C3AR1; C3R1; HNFAG09
UniProt ID: (Human) Q16581, (Rat) O55197, (Mouse) O09047
Entrez Gene ID: (Human) 719, (Rat) 84007, (Mouse) 12267
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.