Abnova
Manufacturer Code:PAB26119
Catalog # 89-127-760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 20 amino acids at extracellular domain of human C3AR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of C3AR1.
For Research Use Only
Protein Aliases: AZ3B; C3a anaphylatoxin chemotactic receptor; C3AR; complement component 3 receptor 1; HNFAG09
Gene Aliases: AZ3B; C3AR; C3AR1; C3R1; HNFAG09
UniProt ID: (Human) Q16581
Entrez Gene ID: (Human) 719
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.