Abnova
Manufacturer Code:PAB29121
Catalog # 89-130-603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Bovine |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Two synthetic peptides (conjugated with KLH) correspoding to Bovine C3 emulsified in Freund's adjuvant. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Chicken polyclonal antibody raised against synthetic peptides of Bovine C3.
For Research Use Only
Protein Aliases: Acylation stimulating protein; ASP; C3-beta-c; C3a anaphylatoxin; C3adesArg; C3bc; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3
Gene Aliases: C3
UniProt ID: (Bovine) Q2UVX4
Entrez Gene ID: (Bovine) 280677
Molecular Function:
complement component
cytokine
defense/immunity protein
enzyme modulator
protease inhibitor
serine protease inhibitor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.