Invitrogen
Manufacturer Code:PA5113295
Catalog # PIPA5113295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human m-AAA protease-interacting protein 1 mitochondrial protein (97-291AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430016H08Rik AA409999 C2orf47 C78988 chromosome 2 open reading frame 47 m-AAA protease-interacting protein 1 mitochondrial Maip1 Matrix AAA peptidase-interacting protein 1 RIKEN cDNA 9430016H08 gene uncharacterized protein C2orf47 homolog mitochondrial uncharacterized protein C2orf47 mitochondrial; m-AAA protease-interacting protein 1, mitochondrial; Matrix AAA peptidase-interacting protein 1; uncharacterized protein C2orf47 homolog, mitochondrial
Gene Aliases: AA409999; C2orf47; C78988; MAIP1
UniProt ID: (Mouse) Q8BHE8, (Human) Q8WWC4
Entrez Gene ID: (Mouse) 68115, (Human) 79568
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.