Abnova
Manufacturer Code:PAB22611
Catalog # 89-124-324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human C22orf32. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant C22orf32.
For Research Use Only
Protein Aliases: DDDD; dJ186O1.1; Essential MCU regulator, mitochondrial; single-pass membrane protein with aspartate-rich tail 1; Single-pass membrane protein with aspartate-rich tail 1, mitochondrial; UPF0466 protein C22orf32, mitochondrial
Gene Aliases: C22orf32; DDDD; dJ186O1.1; EMRE; SMDT1
UniProt ID: (Human) Q9H4I9
Entrez Gene ID: (Human) 91689
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.