Abnova
Manufacturer Code:PAB19695
Catalog # 89-121-222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 201-215 at internal region of human C22orf28. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of C22orf28.
For Research Use Only
Protein Aliases: 3'-phosphate/5'-hydroxy nucleic acid ligase; ankyrin repeat domain 54; DJ149A16.6; focal adhesion-associated protein; HSPC117; RNA-splicing ligase RtcB homolog; RP1-149A16.6; RTCB
Gene Aliases: C22orf28; DJ149A16.6; FAAP; HSPC117; RTCB
UniProt ID: (Human) Q9Y3I0
Entrez Gene ID: (Human) 51493
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.