OriGene
Manufacturer Code:CF504220
Catalog # 50-166-2720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F6 |
Immunogen | Full length human recombit protein of human C20orf3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adipocyte plasma membrane-associated protein; BSCv C20orf3; Protein BSCv; Protein DD16
Gene Aliases: 2310001A20Rik; Ab2-305; AI314817; APMAP; BSCv; C20orf3; C23H20orf3; RGD1308874; UNQ1869/PRO4305
UniProt ID: (Human) Q9HDC9, (Mouse) Q9D7N9, (Rat) Q7TP48
Entrez Gene ID: (Dog) 608089, (Human) 57136, (Mouse) 71881, (Rat) 366227
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.