Proteintech
Manufacturer Code:214931AP150UL
Catalog # 50-173-3448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C20orf160 Fusion Protein Ag15841 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf160 CCM2L; CCM2-like; cerebral cavernous malformation 2-like; Cerebral cavernous malformations 2 protein-like
Gene Aliases: C20orf160; CCM2L; dJ310O13.5; RGD1305202
UniProt ID: (Human) Q9NUG4, (Mouse) Q8VCC6
Entrez Gene ID: (Human) 140706, (Mouse) 228788, (Rat) 311550
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.