R&D Systems
Manufacturer Code:MAB10445SP
Catalog # MAB10445SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 395520 |
Immunogen | Mouse myeloma cell line NS0-derived human CTRP1/C1qTNF1 Arg26-Pro281 Accession &num Q9BXJ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C1q and tumor necrosis factor related protein 1 C1qTNF1 CTRP1G protein-coupled receptor-interacting protein FLJ90694 G protein coupled receptor interacting protein GIPcomplement C1q tumor necrosis factor-related protein 1 ZSIG37; Complement C1q tumor necrosis factor-related protein 1; G protein coupled receptor interacting protein; G protein-coupled receptor-interacting protein; GIP
Gene Aliases: C1QTNF1; CTRP1; GIP; UNQ310/PRO353; ZSIG37
UniProt ID: (Human) Q9BXJ1
Entrez Gene ID: (Human) 114897
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.