Invitrogen
Manufacturer Code:MA140045
Catalog # 01-676-498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 7H8 |
Immunogen | Murine C1q component of the C1 complex. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Complement C1q subcomponent subunit A; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide
Gene Aliases: AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1qg; Ciqc
UniProt ID: (Mouse) P98086, (Mouse) P14106, (Mouse) Q02105
Entrez Gene ID: (Mouse) 12259, (Mouse) 12260, (Mouse) 12262
Molecular Function: cell adhesion molecule extracellular matrix protein extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.