Invitrogen
Manufacturer Code:PA529586
Catalog # PIPA529586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 13 and 225 of Human Complement C1qA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: complement C1q chain A; Complement C1q subcomponent subunit A; complement C1q subcomponent subunit A complement component 1 q subcomponent alpha polypeptide complement component C1q A chain; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component C1q, A chain
Gene Aliases: AI255395; C1q; C1QA
UniProt ID: (Human) P02745, (Mouse) P98086
Entrez Gene ID: (Human) 712, (Mouse) 12259
Molecular Function: cell adhesion molecule extracellular matrix protein extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.