Novus Biologicals
Manufacturer Code:NBP214590
Catalog # NBP214590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: KWCSVRVALYRQRSRTALSKGKDYLVLAQPPLLLPAESVDVSVLQPLSGE LENTNIEREAETVVLGDLSSSMHQDDLVNHTVDAVELEPSHSQTLSQSLL L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf9 CH1 OPT SLP1 SUCO SUN domain containing ossification factor SUN domain containing ossification factor; Membrane protein CH1; osteopotentia; Protein osteopotentia homolog; SUN domain-containing ossification factor; SUN-like protein 1
Gene Aliases: C1orf9; CH1; OPT; SLP1; SUCO
UniProt ID: (Human) Q9UBS9
Entrez Gene ID: (Human) 51430
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.