Proteintech
Manufacturer Code:260741AP150UL
Catalog # 50-173-5194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C1orf9 Fusion Protein Ag23224 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf9 CH1 OPT Protein osteopotentia homolog SUCO; Membrane protein CH1; osteopotentia; protein C1orf9 homolog; protein osteopotentia; Protein osteopotentia homolog; SUN domain-containing ossification factor; SUN-like protein 1
Gene Aliases: 4732491B11; AI848100; AU016931; C1orf9; CH1; OPT; SLP1; SUCO
UniProt ID: (Human) Q9UBS9
Entrez Gene ID: (Human) 51430, (Mouse) 226551
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.