Invitrogen
Manufacturer Code:PA544307
Catalog # PIPA544307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human C1orf77 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromatin target of PRMT1 protein; Friend of PRMT1 protein; Small arginine- and glycine-rich protein; small protein rich in arginine and glycine; SRAG
Gene Aliases: 2500003M10Rik; C1orf77; C3H1orf77; CHTOP; FL-SRAG; FOP; HT031; MNCb-1706; PP7704; SRAG; SRAG-3; SRAG-5
UniProt ID: (Bovine) Q3SYW9, (Human) Q9Y3Y2, (Mouse) Q9CY57, (Rat) Q498T2
Entrez Gene ID: (Bovine) 616638, (Horse) 100056390, (Dog) 480140, (Rabbit) 108175362, (Human) 26097, (Mouse) 66511, (Rat) 361990, (Guinea pig) 100725767
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.