Proteintech
Manufacturer Code:263221AP150UL
Catalog # 50-173-5307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C1orf71 Fusion Protein Ag23732 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf71 CNST Consortin; Consortin; protein phosphatase 1, regulatory subunit 64
Gene Aliases: 9630058J23Rik; C1orf71; CNST; PPP1R64
UniProt ID: (Human) Q6PJW8, (Mouse) Q8CBC4
Entrez Gene ID: (Human) 163882, (Mouse) 226744
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.