Invitrogen
Manufacturer Code:PA555643
Catalog # PIPA555643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human C1orf51 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ChIP-derived repressor of network oscillator; Chrono; circadian associated repressor of transcription; Circadian-associated transcriptional repressor; Computationally highlighted repressor of the network oscillator; hypothetical gene supported by NM_017187; uncharacterized protein C1orf51
Gene Aliases: C1orf51; CHRONO; CIART; GM129; RGD1566380
UniProt ID: (Human) Q8N365, (Mouse) Q3TQ03
Entrez Gene ID: (Human) 148523, (Mouse) 229599, (Rat) 365871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.