Invitrogen
Manufacturer Code:PA543887
Catalog # PIPA543887
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human C1orf51 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ChIP-derived repressor of network oscillator; Chrono; circadian associated repressor of transcription; Circadian-associated transcriptional repressor; Computationally highlighted repressor of the network oscillator; hypothetical gene supported by NM_017187
Gene Aliases: C1orf51; C5H1orf51; CHRONO; CIART; GM129; RGD1566380
UniProt ID: (Human) Q8N365, (Mouse) Q3TQ03
Entrez Gene ID: (Pig) 100156195, (Rabbit) 100341952, (Dog) 100856543, (Horse) 100058902, (Human) 148523, (Mouse) 229599, (Guinea pig) 100720834, (Rat) 365871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.