Abnova
Manufacturer Code:H00388753M03
Catalog # 89-107-790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4A6 |
Immunogen | C1orf31 (NP_001013003.11 a.a. &bsim 125 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant C1orf31.
For Research Use Only
Protein Aliases: Cytochrome c oxidase assembly factor 6 homolog
Gene Aliases: C1orf31; CEMCOX4; COA6
UniProt ID: (Human) Q5JTJ3
Entrez Gene ID: (Human) 388753
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.