Proteintech
Manufacturer Code:242091AP150UL
Catalog # 50-173-4265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C1orf31 Fusion Protein Ag21525 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf31 COA6; Cytochrome c oxidase assembly factor 6 homolog
Gene Aliases: 1810063B05Rik; AI447995; C1orf31; CEMCOX4; COA6
UniProt ID: (Human) Q5JTJ3, (Mouse) Q8BGD8
Entrez Gene ID: (Human) 388753, (Mouse) 67892
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.