Invitrogen
Manufacturer Code:PA5112949
Catalog # PIPA5112949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Lymphocyte expansion molecule protein (1-418AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf177 LEM LEXM Lymphocyte expansion molecule uncharacterized protein C1orf177; Lymphocyte expansion molecule; uncharacterized protein C1orf177; uncharacterized protein C1orf177 homolog
Gene Aliases: C1orf177; LEM; LEXM
UniProt ID: (Human) Q3ZCV2, (Rat) B1H283
Entrez Gene ID: (Human) 163747, (Rat) 500516
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.